Flasks, flat bottom with Quickfit® ground joint
2000 ml, 215 mm, 34/35 NS
Catalog # BIBBFF2L/5S
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, clear, with short neck.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat base allows use with hotplates. Labelling area.
Specifications
- Socket size:34/35 NS
- Capacity:2000 ml
- Height:215 mm
- Seq:24



