Orders LinkContactUsLinkComponent
Dilatometers, Quickfit®
Dilatometer body, with socket size NS 14/23, without stopper
Catalog # BIBBWE12/1
Supplier:   SciLabware
Dilatometers, Quickfit®
Catalog # BIBBWE12/1
Supplier:  SciLabware
Dilatometers, Quickfit®
Dilatometers, Quickfit®
Dilatometer body, with socket size NS 14/23, without stopper
Price
1 170,00 €
Log in for availability
1 * 5 ST
Product Details & Documents
PYREX®, borosilicate glass.
  • Dilatometer body is graduated with an arbitrary scale
  • Interchangable stopper has open top to allow weighting with steel balls etc.
For measuring the expansion of edible oils and fats over the temperature range within which melting takes place.
Ordering information: The body and stopper should be ordered separately.
Specifications
  • Description:
    Dilatometer body, with socket size NS 14/23, without stopper
  • Seq:
    01