Orders LinkContactUsLinkComponent
Column packing support, Quickfit®
Column packing support, Quickfit®
29 mm, 34/35 NS
Catalog # BIBBEX13/25
Supplier:   SciLabware
Column packing support, Quickfit®
Catalog # BIBBEX13/25
Supplier:  SciLabware
Column packing support, Quickfit®
Column packing support, Quickfit®
29 mm, 34/35 NS
Price
92,40 €
Log in for availability
1 * 5 ST
Product Details & Documents
PYREX®, borosilicate glass.
Designed to support Raschig rings within plain fractionating columns. Support is passed through socket of column and rests at the constriction above the cone.
Specifications
  • Cone size:
    34/35 NS
  • Ø:
    29 mm
  • Seq:
    03