Orders LinkContactUsLinkComponent
Splash heads, with sinter, Quickfit®
Splash heads, with sinter, Quickfit®
29/32 NS, 19/26 NS
Catalog # BIBBSH16/19
Supplier:   SciLabware
Splash heads, with sinter, Quickfit®
Catalog # BIBBSH16/19
Supplier:  SciLabware
Splash heads, with sinter, Quickfit®
Splash heads, with sinter, Quickfit®
29/32 NS, 19/26 NS
Price
901,00 €
Log in for availability
1 * 5 ST
Product Details & Documents
PYREX® borosilicate glas
  • Prevents possible contamination of the condenser from 'bumping' solvents
  • Features a coarse porosity 1 sinter to reduce carry over
  • Splash heads feature a 29/32 socket
For use with rotary evaporators.
Specifications
  • Cone size:
    19/26 NS
  • Socket size:
    29/32 NS
  • Seq:
    01