Orders LinkContactUsLinkComponent
Stopcocks, Rotaflo®
6 mm, 13 mm
  BIBBHP6ST/10
 :   SciLabware
Stopcocks, Rotaflo®
  BIBBHP6ST/10
Stopcocks, Rotaflo®
Stopcocks, Rotaflo®
6 mm, 13 mm
587,00 €
1 * 5 ST
PYREX®, borosilicate glass with PTFE needle valve.
  • Lubricant is not required (no seizure, no contamination with grease)
  • Interchangeable elements
  • Precise flow control
These chemically inert stopcocks can be used at temperatures up to +200 °C. Two different models are available: general purpose (vacuum rating up to 10-4 mm Hg) and high performance (vacuum rating up to 10-9 mm Hg).
  • Hole Ø:
    6 mm
  • Seq:
    12
  • For tube I-Ø:
    13 mm
Accessories
  •  
    Description
    For
  • 229-0230
    Replacement key, bore Ø 6 mm
    High performance Rotaflo® stopcocks
    1 * 2 ST - 124,00 €
  •  
    Description
    For
  • 229-0231
    Replacement key, bore Ø 6 mm
    General purpose Rotaflo® stopcocks
    1 * 5 ST - 171,00 €
  •  
    Description
    For
  • BIBBLR6
    Replacement locking ring for key with bore 6 mm
    Rotaflo® stopcocks
    1 * 5 ST - 40,90 €
  •  
    Description
    For
  • 229-0226
    Replacement key, bore Ø 10 mm
    High performance Rotaflo® stopcocks
    1 * 1 ST - 79,50 €
  •  
    Description
    For
  • 229-0229
    Replacement key, bore Ø 3 mm
    High performance Rotaflo® stopcocks
    1 * 2 ST - 88,70 €
  •  
    Description
    For
  • BIBBLR3
    Replacement locking ring for key with bore 3 mm
    Rotaflo® stopcocks
    1 * 5 ST - 27,90 €
  •  
    Description
    For
  • 229-0234
    Replacement key, bore Ø 10 mm
    General purpose Rotaflo® stopcocks
    1 * 5 ST - 266,00 €
  •  
    Description
    For
  • BIBBGP3K
    Replacement key, bore Ø 3 mm
    General purpose Rotaflo® stopcocks
    1 * 5 ST - 110,00 €
  •  
    Description
    For
  • BIBBLR10
    Replacement locking ring for key with bore 10 mm
    Rotaflo® stopcocks
    1 * 5 ST - 38,20 €