Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH Buffers

pH Buffers

pH buffer, 3,9 M NaCl, 4,010, 250 ml, 250 mL
Catalog # METN52004103
Supplier:   Mettler - Toledo
pH Buffers
Catalog # METN52004103
Supplier:  Mettler - Toledo
CAS Number:  
pH Buffers

pH buffer, 3,9 M NaCl, 4,010, 250 ml, 250 mL

CAS Number:  
Price
70,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    pH buffer, 3,9 M NaCl
  • Pk Info:
    250 ml
  • pH value:
    4,010

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.