Order Entry
Belgium
Orders LinkContactUsLinkComponent
Corning® CoolCell® containers

Corning® CoolCell® containers

CoolCell® SV2, exposed tops, 12 standard 2.0 mL injectable ampules, Purple
  479-1918
   Safer Human and Environmental Health
 :   Corning
Corning® CoolCell® containers
  479-1918
 :  Corning
   Safer Human and Environmental Health
Corning® CoolCell® containers
Corning® CoolCell® containers

CoolCell® SV2, exposed tops, 12 standard 2.0 mL injectable ampules, Purple

 
386,00 €
   
1 * 1 ST
 

CoolCell® alcohol-free cell freezing containers ensure standardised controlled-rate −1 °C/minute cell freezing in a −80 °C freezer, without alcohol or any fluids.

  • No alcohol. no fluids, no pre-cooling
  • No variability, all vials have uniform freeze rate
  • Radially symmetric design ensures vial consistency
  • No on-going cost, no alcohol purchase or disposal
  • No stuck lids, ergonomic lid comes off easily when frozen
  • Not cold to the touch when removing from the −80 °C freezer
  • Quick re-use time, ready to use again after five minutes

Proven for use with a variety of cell types including stem cells, primary cells, PBMC cell lines, insect cells, yeast and others. The patent-pending CoolCell® technology utilises a thermo-conductive alloy core and highly-insulative outer material to control the rate of heat removal and provide reproducible cell cryopreservation. CoolCell units are easy to use and deliver comparable results to expensive programmable freezers.

CoolCell® LX
For 12 standard 1,0 to 2,0 ml cryogenic vials, 1,0 ml fill per vial. Radially symmetric for uniform vial freezing. Numbered wells for easy sample identification. Beveled lid for secure gripping and easy opening. Exposed vial tops when lid is open for quick, organised removal of frozen samples.
CoolCell® FTS30
For 30 standard 1,0 to 2,0 ml cryogenic vials, 1,0 ml fill per vial. Controlled micro-convection for uniform freezing of 30 vials. Removable vial tray for one-step transfer of samples into and out of freezing chamber.

CoolCell® 5 mL LX
For 12 standard 3,5 to 5,0 ml fill cryogenic vials, 3,5 to 5,0 fill per vial. Radially symmetric for uniform vial freezing. Numbered wells for easy sample identification. Beveled lid for secure gripping and easy opening. Exposed vial tops when lid is open for quick, organised removal of frozen samples.
CoolCell® SV2 and CoolCell® SV10
For 12 standard 2,0 ml injectable ampules, 1,0 ml fill per ampule (SV2). For 6 standard 10,0 ml injectable ampules, 5,0 ml fill per ampule (SV10). Radially symmetric for uniform freezing of injectable ampules. Easy open lid. Exposed vial tops when lid is open for quick, organized removal of frozen samples.

CoolCell SV10 Stem Cell Cryopreservation System
includes the XT Starter Benchtop Cooler, Corning® CoolRack SV10 and CoolCell SV10 Cell Freezing Container for 6 injectable vials.
CoolCell® Filler Vials
To ensure cell freezing rate consistency and uniform results when using CoolCell containers, insert a CoolCell Filler Vial into empty wells when freezing less than a full load. Suitable for repeated use and compatible with CoolCell LX, CoolCell FTS30 and CoolCell 5 ml LX containers.
CoolCell® FTS30 Vial Module
FTS30 Vial Module is a holder for 30×1,0 ml or 2,0 ml cryogenic vials that allows one-step insertion and removal of all 30 vials at once. Fits into a standard 5,0×5,0×2,0" cryostorage box. Compatible with dry ice and liquid nitrogen.

Documents

 
  • For:
    12 standard 2.0 mL injectable ampules
  • Colour:
    Purple
  • Description:
    CoolCell® SV2, exposed tops
  • Seq:
    12
  • Cat. No.:
    479-1918
  • Environmentally Preferable:
    Safer Human and Environmental Health
Accessories

 

  •  
    Description
    For
     
     
  • 479-1961
    CoolCell® 2 mL filler vials
    Use with CoolCell LX and CoolCell FTS30 containers
       
    1 * 6 ST - 29,20 €

 

  •  
    Description
     
     
  • 479-1846
    Cryogenic vial grippers, assorted colours
       
    1 * 5 ST - 56,00 €

 

  •  
    Description
    For
     
     
  • 479-1962
    CoolCell® 5 mL filler vials
    Use with CoolCell 5mL LX container
       
    1 * 6 ST - 29,20 €

 

  •  
    Description
    For
     
     
  • 479-1852
    Removable cryogenic vial module, holds 30 cryogenic vials (1.0 mL or 2.0 mL)
    CoolCell FTS30
       
    1 * 10 ST - 121,00 €