Order Entry
Belgium
Orders LinkContactUsLinkComponent
Corning® CoolCell® containers

Corning® CoolCell® containers

CoolCell® LX, exposed tops, 12 standard 1,0 mL to 2,0 mL cryogenic vials, Green
Catalog # 479-1842
   Safer Human and Environmental Health
Supplier:   Corning
Corning® CoolCell® containers
Catalog # 479-1842
Supplier:  Corning
   Safer Human and Environmental Health
Corning® CoolCell® containers
Corning® CoolCell® containers

CoolCell® LX, exposed tops, 12 standard 1,0 mL to 2,0 mL cryogenic vials, Green

Price
202,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

CoolCell® alcohol-free cell freezing containers ensure standardised controlled-rate −1 °C/minute cell freezing in a −80 °C freezer, without alcohol or any fluids.

  • No alcohol. no fluids, no pre-cooling
  • No variability, all vials have uniform freeze rate
  • Radially symmetric design ensures vial consistency
  • No on-going cost, no alcohol purchase or disposal
  • No stuck lids, ergonomic lid comes off easily when frozen
  • Not cold to the touch when removing from the −80 °C freezer
  • Quick re-use time, ready to use again after five minutes

Proven for use with a variety of cell types including stem cells, primary cells, PBMC cell lines, insect cells, yeast and others. The patent-pending CoolCell® technology utilises a thermo-conductive alloy core and highly-insulative outer material to control the rate of heat removal and provide reproducible cell cryopreservation. CoolCell units are easy to use and deliver comparable results to expensive programmable freezers.

CoolCell® LX
For 12 standard 1,0 to 2,0 ml cryogenic vials, 1,0 ml fill per vial. Radially symmetric for uniform vial freezing. Numbered wells for easy sample identification. Beveled lid for secure gripping and easy opening. Exposed vial tops when lid is open for quick, organised removal of frozen samples.
CoolCell® FTS30
For 30 standard 1,0 to 2,0 ml cryogenic vials, 1,0 ml fill per vial. Controlled micro-convection for uniform freezing of 30 vials. Removable vial tray for one-step transfer of samples into and out of freezing chamber.

CoolCell® 5 mL LX
For 12 standard 3,5 to 5,0 ml fill cryogenic vials, 3,5 to 5,0 fill per vial. Radially symmetric for uniform vial freezing. Numbered wells for easy sample identification. Beveled lid for secure gripping and easy opening. Exposed vial tops when lid is open for quick, organised removal of frozen samples.
CoolCell® SV2 and CoolCell® SV10
For 12 standard 2,0 ml injectable ampules, 1,0 ml fill per ampule (SV2). For 6 standard 10,0 ml injectable ampules, 5,0 ml fill per ampule (SV10). Radially symmetric for uniform freezing of injectable ampules. Easy open lid. Exposed vial tops when lid is open for quick, organized removal of frozen samples.

CoolCell SV10 Stem Cell Cryopreservation System
includes the XT Starter Benchtop Cooler, Corning® CoolRack SV10 and CoolCell SV10 Cell Freezing Container for 6 injectable vials.
CoolCell® Filler Vials
To ensure cell freezing rate consistency and uniform results when using CoolCell containers, insert a CoolCell Filler Vial into empty wells when freezing less than a full load. Suitable for repeated use and compatible with CoolCell LX, CoolCell FTS30 and CoolCell 5 ml LX containers.
CoolCell® FTS30 Vial Module
FTS30 Vial Module is a holder for 30×1,0 ml or 2,0 ml cryogenic vials that allows one-step insertion and removal of all 30 vials at once. Fits into a standard 5,0×5,0×2,0" cryostorage box. Compatible with dry ice and liquid nitrogen.

Documents

Specifications
  • For:
    12 standard 1,0 mL to 2,0 mL cryogenic vials
  • Colour:
    Green
  • Description:
    CoolCell® LX, exposed tops
  • Seq:
    03
  • Cat. No.:
    479-1842
  • Environmentally Preferable:
    Safer Human and Environmental Health
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • CoolCell® 2 mL filler vials
    CoolCell® 2 mL filler vials
    479-1961
    Use with CoolCell LX and CoolCell FTS30 containers
    Log in for availability
    1 * 6 ST - 29,20 €

Product Information

  • Catalog #
    Availability
    Price
  • Cryogenic vial grippers, assorted colours
    Cryogenic vial grippers, assorted colours
    479-1846
    Log in for availability
    1 * 5 ST - 56,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • CoolCell® 5 mL filler vials
    CoolCell® 5 mL filler vials
    479-1962
    Use with CoolCell 5mL LX container
    Log in for availability
    1 * 6 ST - 29,20 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Removable cryogenic vial module, holds 30 cryogenic vials (1.0 mL or 2.0 mL)
    Removable cryogenic vial module, holds 30 cryogenic vials (1.0 mL or 2.0 mL)
    479-1852
    CoolCell FTS30
    Log in for availability
    1 * 10 ST - 121,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.