ContactUsLinkComponent
LDPE bags, clear, compatible for use in cleanroom. Thickness 2mil (50µm), Width 20 (508mm), Length 48 (1219mm). Cleanliness level 100 as per NASA JPAG 5322.1, NASA KSC-C-123J, IEST-CC-1246D. 100 pcs per double-pack 1 * 100 items
Catalog # FISHPCII20X48X2MIL
Supplier:  FISHER CONTAINER
LDPE bags, clear, compatible for use in cleanroom. Thickness 2mil (50µm), Width 20 (508mm), Length 48 (1219mm). Cleanliness level 100 as per NASA JPAG 5322.1, NASA KSC-C-123J, IEST-CC-1246D. 100 pcs per double-pack 1 * 100 items
Catalog # FISHPCII20X48X2MIL
Supplier:  FISHER CONTAINER