Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Worktables

Stainless steel, 2000×700×900 mm
  HUPF75R60302-2000
Worktables
  HUPF75R60302-2000
Worktables
Worktables

Stainless steel, 2000×700×900 mm

 
3 620,00 €
   
1 * 1 ST
 

This open version work table features closed table top and bottom side covered with 0,8 mm sheet steel.

  • Particle board stuffing
  • Sandwich panelled table top

Working table made of stainless steel with grain size 400 ground and material thickness of 1 mm.

 
  • Material:
    Stainless steel
  • Seq:
    03
  • W×D×H:
    2000×700×900 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.