Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Krebs-Ringer solution, bicarbonate-buffered

Krebs-Ringer Solution, bicarbonate-buffered, 1 L
Catalog # J67591.K2
Krebs-Ringer solution, bicarbonate-buffered
Catalog # J67591.K2
CAS Number:  
Krebs-Ringer solution, bicarbonate-buffered
Krebs-Ringer solution, bicarbonate-buffered

Krebs-Ringer Solution, bicarbonate-buffered, 1 L

CAS Number:  
Price
181,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Contains: 120 mM sodium chloride, 5 mM potassium chloride, 2 mM calcium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, and 5,5 mM D-glucose.

  • pH 7,3
  • Filtered through 0,22 micron filter
  • Prepared in 18,2 megohms water.

Physical Form: Liquid
Appearance: Colourless

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Krebs-Ringer Solution, bicarbonate-buffered
  • Cat. No.:
    J67591.K2
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.