Krebs-Ringer solution, bicarbonate-buffered
Krebs-Ringer Solution, bicarbonate-buffered, 500 mL
Supplier: Thermo Fisher Scientific
Krebs-Ringer solution, bicarbonate-buffered
Catalog # J67591.AP
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Contains: 120 mM sodium chloride, 5 mM potassium chloride, 2 mM calcium chloride, 1 mM magnesium chloride, 25 mM sodium bicarbonate, and 5,5 mM D-glucose.
- pH 7,3
- Filtered through 0,22 micron filter
- Prepared in 18,2 megohms water.
Physical Form: Liquid
Appearance: Colourless
Documents
Specifications
Specifications
- Pk:500 mL
- Description:Krebs-Ringer Solution, bicarbonate-buffered
- Cat. No.:J67591.AP
- Physical form:Liquid
- Appearance:Colourless
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



