ContactUsLinkComponent
Caps for screw thread microtubes ********************************** Made of polypropylene. Feature an internal O-ring to ensure leakproof seal Colur: Neutral Dimensions 13.0 x 6.0 mm. 1 * 500 items
Catalog # DELT409112/0
Supplier:  DELTALAB
Caps for screw thread microtubes ********************************** Made of polypropylene. Feature an internal O-ring to ensure leakproof seal Colur: Neutral Dimensions 13.0 x 6.0 mm. 1 * 500 items
Catalog # DELT409112/0
Supplier:  DELTALAB