Maintenance-free pH sensor for harsh chemical environments, Cleaning: SIP, Autoclavation, no cable, Connector head: Memosens, Integr ated temperature sensor: NTC 30 kOhm, Diaphragm: 2 x Single Pore, Material: glass shaft, Electrolyte: Polisolve Plus 1 * 1 items
Catalog # HAMI242428-3413
Supplier: HAMILTON BONADUZ
Frequently Bought Together