CuVer 1 Copper Reagent Powder Pillows, for 10 ml sample, pk/1000 MR 0.04 - 5.00 mg/L Cu 1 * 1.000 items
Catalog # HACH2105828
Supplier: Hach
CuVer 1 Copper Reagent Powder Pillows, for 10 ml sample, pk/1000 MR 0.04 - 5.00 mg/L Cu 1 * 1.000 items
Catalog # HACH2105828
Supplier: Hach
Frequently Bought Together
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals