Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Test kits, drop count tests

Sodium thiosulfate standard solution, stabilised, 0,00246 N, 500 ml bottle, 500 mL
Catalog # HACH2408549
Supplier:   Hach
Test kits, drop count tests
Catalog # HACH2408549
Supplier:  Hach
CAS Number:  
Test kits, drop count tests
Test kits, drop count tests

Sodium thiosulfate standard solution, stabilised, 0,00246 N, 500 ml bottle, 500 mL

CAS Number:  
Price
49,80 €
Log in for availability
1 * 500 mL
Product Details & Documents

The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.

  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit

 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Sodium thiosulfate standard solution, stabilised, 0,00246 N, 500 ml bottle

Specifications

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.