Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH buffer solutions

pH buffer standard solution with certificate of analysis, 125 ml, 7,00, 125 mL
Catalog # HACHLZW9461.98
Supplier:   Hach
pH buffer solutions
Catalog # HACHLZW9461.98
Supplier:  Hach
CAS Number:  
pH buffer solutions
pH buffer solutions

pH buffer standard solution with certificate of analysis, 125 ml, 7,00, 125 mL

CAS Number:  
Price
16,10 €
Log in for availability
1 * 125 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    125 mL
  • Description:
    pH buffer standard solution with certificate of analysis, 125 ml
  • pH value:
    7,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.