pH buffer solutions, colour coded
Buffer solution, 7,00, ±0,02, Phosphate, Yellow, 25 °C, 15 mL
HACH2283536
: Hach
For calibration of pH meters
- Colour coded for easy identification
- Accuracy (pH): ±0,02 at 25 °C
: Traceable to NIST.
- pH buffer type:Phosphate
- Pk:15 mL
- Reference temperature:25 °C
- Colour:Yellow
- Description:Buffer solution
- pH accuracy:±0,02
- pH value:7,00



