pH buffer solutions, colour coded
Buffer solution, 4,01, ±0,02, Phthalate, Red, 25 °C, 500 mL
Supplier: Hach
Jump to:
Product Details & Documents
For calibration of pH meters
- Colour coded for easy identification
- Accuracy (pH): ±0,02 at 25 °C
Certifications: Traceable to NIST.
Specifications
Specifications
- pH buffer type:Phthalate
- Pk:500 mL
- Reference temperature:25 °C
- Colour:Red
- Description:Buffer solution
- pH accuracy:±0,02
- pH value:4,01



