Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH buffer solutions, colour coded

pH buffer solutions, colour coded

Buffer solution, 4,01, ±0,02, Phthalate, Red, 25 °C, 20 L
Catalog # HACH2283461
Supplier:   Hach
pH buffer solutions, colour coded
Catalog # HACH2283461
Supplier:  Hach
CAS Number:  
pH buffer solutions, colour coded
pH buffer solutions, colour coded

Buffer solution, 4,01, ±0,02, Phthalate, Red, 25 °C, 20 L

CAS Number:  
Price
235,00 €
Log in for availability
1 * 20 L
Product Details & Documents

For calibration of pH meters

  • Colour coded for easy identification
  • Accuracy (pH): ±0,02 at 25 °C

Certifications: Traceable to NIST.

Specifications

Specifications

  • pH buffer type:
    Phthalate
  • Pk:
    20 L
  • Reference temperature:
    25 °C
  • Colour:
    Red
  • Description:
    Buffer solution
  • pH accuracy:
    ±0,02
  • pH value:
    4,01

Specifications