Radial fan with flange DN 75, 230 V, 50 Hz 2800 min-1, 62 Hz 3370 min-1, with capacitor 0,08 kW, IP 55, housing pps 1 * 1 items
Catalog # KOTT851-00052
Supplier: KOETTERMANN
Radial fan with flange DN 75, 230 V, 50 Hz 2800 min-1, 62 Hz 3370 min-1, with capacitor 0,08 kW, IP 55, housing pps 1 * 1 items
Catalog # KOTT851-00052
Supplier: KOETTERMANN
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK