α-Cyclodextrin, white powder
5 g
Catalog # ICNA0219005205
Supplier: MP Biomedicals
α-Cyclodextrin, white powder
Catalog # ICNA0219005205
Supplier: MP Biomedicals
CAS Number: 10016-20-3
Jump to:
Product Details & Documents
α-Cyclodextrin is reported useful for the selective precipitation of enantiomeric, positional or structural isomers. Cyclodextrins have hydrophobic cavities; form inclusion compounds with organic substances, salts and halogens in the solid state or in aqueous solutions.
Soluble in water (50 mg/ml).
- CAS Number:10016-20-3
- Formula:C₃₆H₆₀O₃₀
- MDL Number:MFCD00078207
- Molecular Weight:972.84 g/mol
- Melting Pt:278 °C (lit.)
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Identity Test:Passes
- Loss on Drying:≤13%
- Optical Rotation:+148 ± 5°(c=1, water)
Specifications
- Pk:5 g



