Order Entry
Belgium
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB), liquid substrate

3,3',5,5'-Tetramethylbenzidine (TMB), liquid substrate

50 mL
Catalog # ICNA0215234650
Irritant
Supplier:   MP Biomedicals
3,3',5,5'-Tetramethylbenzidine (TMB), liquid substrate
Catalog # ICNA0215234650
Supplier:  MP Biomedicals
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB), liquid substrate

50 mL

CAS Number:  54827-17-7
Price
81,00 €
Log in for availability
1 * 50 mL
Product Details & Documents

TMB is a chromogenic substrate for Horse Radish Peroxidase (HRP). MP Biolomedicals TMB Solution is a convenient, non toxic and ready to use stable chromogen solution suitable for peroxidase and pseudo-peroxidase reactions in ELISA, it provides consistent performance and sensitivity to deliver reproducible results

  • Ready-to-Use, convenient
  • Reproducible Results
  • Consistent Performance and Sensitivity
  • Stabilized chromogen solution
  • For ELISA procedures and peroxidase and pseudo-peroxidase reactions
  • Stable up to 24 months
  • Non toxic.

Metabolism of the TMB substrate by HRP creates a soluble bluish reaction product that may be quantified by measuring absorbance at 370 or 665 nm. Reaction can be stopped by addition of 1 volume of 1N hydrocloric or sulfurc acid to yield a yellow reaction product that can be measured by absorbance at 450 nm (within one hour).

Caution: Research Use Only (RUO).

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Functionality Test :
    Passes
  • pH:
    3.1 ± 0.5
  • Expiration Date:
    Month/Day/Year

Specifications

  • Pk:
    50 mL

Specifications