Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bromochlorophenol blue, powder

Bromochlorophenol blue, powder

Glass bottle for solids, 25 g
Catalog # ICNA0210112325
Supplier:   MP Biomedicals
Bromochlorophenol blue, powder
Catalog # ICNA0210112325
Supplier:  MP Biomedicals
CAS Number:  2553-71-1
Bromochlorophenol blue, powder

Glass bottle for solids, 25 g

CAS Number:  2553-71-1
Price
87,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Bromophenol blue is a tracking dye for alkaline and neutral buffer systems. It is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.

Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.

Bromophenol blue is a tracking dye for alkaline and neutral buffer systems. It is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.

Soluble in ethanol (0,1% w/v - clear yellow solution), methanol (40 mg/4 ml), EGME, water, benzene.

Synonyms:  4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromo-6-chlorophenol] S,S-dioxide
  • CAS Number:2553-71-1
  • Formula:C₁₉H₁₀Br₂Cl₂O₅S
  • MDL Number:MFCD00047291
  • Molecular Weight:581.06 g/mol
  • Boiling Pt:603 °C (1013 hPa)
  • Melting Pt:273 °C
  • EINECS:219-861-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.