Order Entry
Belgium
Orders LinkContactUsLinkComponent
Lecithin

Lecithin

0,45 kg
N° de catalogue ICNA0210214801
Fournisseur:   MP Biomedicals
Lecithin
N° de catalogue ICNA0210214801
Fournisseur:  MP Biomedicals
Numéro CAS:  8002-43-5
Lecithin

0,45 kg

Numéro CAS:  8002-43-5
Prix
57,00 €
Log in for availability
1 * 0,45 kg
Product Details & Documents

Lecithin is a good source of choline, which may help with brain function and growth. Used as an emulsifier and solubilizer for drugs in an aqueous solution, also as a vehicle for liposomes. Lecithin is a phosphatidylcholine of soybean origin.

Storage temperature: -20 °C

  • CAS Number:8002-43-5
  • MDL Number:MFCD00147406
Specifications

Specifications

  • Pk:
    0,45 kg

Specifications