Order Entry
Belgium
Orders LinkContactUsLinkComponent
Polysorbate 20, liquid

Polysorbate 20, liquid

100 mL
Catalog # ICNA0210316880
Supplier:   MP Biomedicals
Polysorbate 20, liquid
Catalog # ICNA0210316880
Supplier:  MP Biomedicals
CAS Number:  9005-64-5
Polysorbate 20, liquid
Polysorbate 20, liquid

100 mL

CAS Number:  9005-64-5
Price
32,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Polysorbate 20 is nonionic surfactant derived from sorbitan ester.

  • Refractive Index: n20/D 1.468(Lit.)
  • Viscosity: 400 cps at 25 °C.(Lit.)
  • Critical Micelle Concentration (CMC): 0.06mM

Polysorbate 20 is typically used as an emulsifying agent for the preparation of stable oil-in-water emulsions, particularly in pharmaceutical applications. It has been used in pre-extraction of membranes to remove peripheral proteins. It has been used as a blocking agent on nitrocellulose.

Caution: Research Use Only (RUO).

Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C (lit.)
  • Density:1.095 g/mL (lit.)
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Identity Test A:
    Passes
  • Identity Test B:
    Passes
  • FTIR:
    Conforms to Reference
  • Solubility (1g/20mL):
    Clear, Foamy Solution
  • Specific Gravity:
    1.08-1.113
  • Water Content:
    ≤3%
  • Saponification Number:
    40-50

Specifications

  • Pk:
    100 mL

Specifications