Orders LinkContactUsLinkComponent
Sterilisation Boxes for Pipettes
Pipette box for the storage of pipettes (315 - 485 mm)
Catalog # BRND28935
Supplier:   Brand
Sterilisation Boxes for Pipettes
Catalog # BRND28935
Supplier:  Brand
Sterilisation Boxes for Pipettes
Sterilisation Boxes for Pipettes
Pipette box for the storage of pipettes (315 - 485 mm)
Price
81,50 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These pipette boxes made of aluminum are ideal for the storage and sterilisation of pipettes with different lengths.
  • With inner silicone coating
  • Very good temperature resistance
  • Very good chemical resistance
Spécifications
  • Description:
    Pipette box for the storage of pipettes (315 - 485 mm)
  • Seq:
    04