Sterilisation Boxes for Pipettes
Pipette box for the storage of pipettes (315 - 385 mm)
Catalog # BRND28930
Supplier: Brand
Aller à :
Product Details & Documents
These pipette boxes made of aluminum are ideal for the storage and sterilisation of pipettes with different lengths.
- With inner silicone coating
- Very good temperature resistance
- Very good chemical resistance
Spécifications
- Description:Pipette box for the storage of pipettes (315 - 385 mm)
- Seq:03