Order Entry
Belgium
Orders LinkContactUsLinkComponent
Shaking water baths, Precision™

Shaking water baths, Precision™

Dubnoff shaking water bath, 15 L, Precision™ DUB 15
Catalog # HAKATSDUB15
Supplier:   THERMO ELECTRON LED
Shaking water baths, Precision™
Catalog # HAKATSDUB15
Shaking water baths, Precision™
Shaking water baths, Precision™

Dubnoff shaking water bath, 15 L, Precision™ DUB 15

Price
4 120,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Precision™ water baths support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culturing and genetics sequencing.

  • Coil-free interior for easy cleaning
  • Four temperature and shaking speed presets
  • Audible alarms protect work
  • Auto-on and auto-off detectors
  • Low-fluid detection
  • Monitor with icon based graphic display

The shallow form shaking bath has a removable tray that allows it to be used with smaller sample containers. Dubnoff shaking water baths are designed specifically for applications that require samples to be incubated in a controlled atmosphere.

Delivery information: Supplied with one large and two small gassing hoods, stainless steel gable cover and shaking tray. Please note flask clips and fasteners must be ordered separately.

Documents

Specifications
  • Description:
    Dubnoff shaking water bath, 15 L, Precision™ DUB 15
  • Seq:
    02
  • Cat. No.:
    HAKATSDUB15
  • Temperature uniformity:
    ±0,5 °C
  • Model:
    Precision™ DUB 15
  • W×D×H:
    636×396×250 mm
  • Temperature range:
    5...100 °C
  • Temperature stability:
    ±0,1 %
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • Clip fastener
    Clip fastener
    HAKA3166189
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 25 ST - 151,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Test tube tray, 13 - 25 mm, 127×259 mm
    Test tube tray, 13 - 25 mm, 127×259 mm
    HAKA3161597
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 886,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask tray, 25 ml, 127×259 mm
    Flask tray, 25 ml, 127×259 mm
    HAKA3161599
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 907,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Large gassing hood, 285×288 mm
    Large gassing hood, 285×288 mm
    HAKA3162640
    TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 403,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • High wall tray, large, 285,75×609,6×190,5 mm
    High wall tray, large, 285,75×609,6×190,5 mm
    HAKA3164717
    TSSWB27
    Log in for availability
    1 * 1 ST - 712,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 50 ml flasks
    Flask clip (without clip fastener), for 50 ml flasks
    HAKA3166198
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 98,80 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 125 ml flasks
    Flask clip (without clip fastener), for 125 ml flasks
    HAKA3166221
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 79,70 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 25 ml flasks
    Flask clip (without clip fastener), for 25 ml flasks
    HAKA3166227
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 79,70 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Small gassing hood, 142×288 mm
    Small gassing hood, 142×288 mm
    HAKA3162639
    TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 283,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • High wall tray, small, 285,75×317,5×190,5 mm
    High wall tray, small, 285,75×317,5×190,5 mm
    HAKA3164716
    TSSWB15, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 600,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Microfuge tube rack, 0,5 ml, 127×101×39 mm
    Microfuge tube rack, 0,5 ml, 127×101×39 mm
    HAKA3166184
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 327,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Microfuge tube rack, 1 ml, 127×101×39 mm
    Microfuge tube rack, 1 ml, 127×101×39 mm
    HAKA3166185
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 311,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 500 ml flasks
    Flask clip (without clip fastener), for 500 ml flasks
    HAKA3166199
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 79,70 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Test tube clips, for 13 - 25 mm tubes
    Test tube clips, for 13 - 25 mm tubes
    HAKA3166216
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 101,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 1000 ml flasks
    Flask clip (without clip fastener), for 1000 ml flasks
    HAKA3166200
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 222,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 250 ml flasks
    Flask clip (without clip fastener), for 250 ml flasks
    HAKA3166566
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 76,10 €