Order Entry
Belgium
Orders LinkContactUsLinkComponent
Shaking water baths, Precision™

Shaking water baths, Precision™

Shaking water bath, 15 L, Precision™ SWB 15
Catalog # HAKATSSWB15
Supplier:   THERMO ELECTRON LED
Shaking water baths, Precision™
Catalog # HAKATSSWB15
Shaking water baths, Precision™
Shaking water baths, Precision™

Shaking water bath, 15 L, Precision™ SWB 15

Price
3 030,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Precision™ water baths support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culturing and genetics sequencing.

  • Coil-free interior for easy cleaning
  • Four temperature and shaking speed presets
  • Audible alarms protect work
  • Auto-on and auto-off detectors
  • Low-fluid detection
  • Monitor with icon based graphic display

The shallow form shaking bath has a removable tray that allows it to be used with smaller sample containers. Dubnoff shaking water baths are designed specifically for applications that require samples to be incubated in a controlled atmosphere.

Delivery information: Supplied with one large and two small gassing hoods, stainless steel gable cover and shaking tray. Please note flask clips and fasteners must be ordered separately.

Documents

Specifications
  • Description:
    Shaking water bath, 15 L, Precision™ SWB 15
  • Seq:
    01
  • Cat. No.:
    HAKATSSWB15
  • Temperature uniformity:
    ±0,5 °C
  • Model:
    Precision™ SWB 15
  • W×D×H:
    632×394×249 mm
  • Temperature range:
    5...100 °C
  • Temperature stability:
    ±0,1 %
Accessories

Product Information

  • Catalog #
    For
    Availability
    Price
  • Clip fastener
    Clip fastener
    HAKA3166189
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 25 ST - 151,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Test tube tray, 13 - 25 mm, 127×259 mm
    Test tube tray, 13 - 25 mm, 127×259 mm
    HAKA3161597
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 886,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask tray, 25 ml, 127×259 mm
    Flask tray, 25 ml, 127×259 mm
    HAKA3161599
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 907,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Large gassing hood, 285×288 mm
    Large gassing hood, 285×288 mm
    HAKA3162640
    TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 403,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • High wall tray, large, 285,75×609,6×190,5 mm
    High wall tray, large, 285,75×609,6×190,5 mm
    HAKA3164717
    TSSWB27
    Log in for availability
    1 * 1 ST - 712,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 50 ml flasks
    Flask clip (without clip fastener), for 50 ml flasks
    HAKA3166198
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 98,80 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 125 ml flasks
    Flask clip (without clip fastener), for 125 ml flasks
    HAKA3166221
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 79,70 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 25 ml flasks
    Flask clip (without clip fastener), for 25 ml flasks
    HAKA3166227
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 79,70 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Small gassing hood, 142×288 mm
    Small gassing hood, 142×288 mm
    HAKA3162639
    TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 283,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • High wall tray, small, 285,75×317,5×190,5 mm
    High wall tray, small, 285,75×317,5×190,5 mm
    HAKA3164716
    TSSWB15, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 600,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Microfuge tube rack, 0,5 ml, 127×101×39 mm
    Microfuge tube rack, 0,5 ml, 127×101×39 mm
    HAKA3166184
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 327,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Microfuge tube rack, 1 ml, 127×101×39 mm
    Microfuge tube rack, 1 ml, 127×101×39 mm
    HAKA3166185
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 311,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 500 ml flasks
    Flask clip (without clip fastener), for 500 ml flasks
    HAKA3166199
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 79,70 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Test tube clips, for 13 - 25 mm tubes
    Test tube clips, for 13 - 25 mm tubes
    HAKA3166216
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 101,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 1000 ml flasks
    Flask clip (without clip fastener), for 1000 ml flasks
    HAKA3166200
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 222,00 €

Product Information

  • Catalog #
    For
    Availability
    Price
  • Flask clip (without clip fastener), for 250 ml flasks
    Flask clip (without clip fastener), for 250 ml flasks
    HAKA3166566
    TSSWB15, TSSWB27, TSSWB15S, TSDUB15
    Log in for availability
    1 * 1 ST - 76,10 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.