ContactUsLinkComponent
Cylinders graduated,VOLAC,USP,5ml:0.1,class A,ISO/ASTM,Boro,hexag.feet,spouted,individually tested,certified and serially numbered 1 * 2 items
Catalog # POULUS263/WAC/A/2
Supplier:  Poulten & Graf
undefined
Cylinders graduated,VOLAC,USP,5ml:0.1,class A,ISO/ASTM,Boro,hexag.feet,spouted,individually tested,certified and serially numbered 1 * 2 items
Catalog # POULUS263/WAC/A/2
Supplier:  Poulten & Graf
Frequently Bought Together