ContactUsLinkComponent
Cell changer Peltier-heated 6 positions, 10mm pathlength (requires Temperature control unit 80-2105-49) 1 * 1 items
Catalog # SCLI80-2106-04
Supplier:  BIOCHROM
Cell changer Peltier-heated 6 positions, 10mm pathlength (requires Temperature control unit 80-2105-49) 1 * 1 items
Catalog # SCLI80-2106-04
Supplier:  BIOCHROM
Frequently Bought Together