Orders LinkContactUsLinkComponent
Cables
Cables
InLab® cable S7-no connector, 3 m
Catalog # 661-0268
Supplier:   Mettler - Toledo
Cables
Catalog # 661-0268
Supplier:  Mettler - Toledo
Cables
Cables
InLab® cable S7-no connector, 3 m
Price
82,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Documents
Specifications
  • Description:
    InLab® cable S7-no connector
  • Length:
    3 m
  • Seq:
    169