Cables
InLab® cable S7-DIN, 1,2 m
Catalog # 661-0261
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Specifications
- Description:InLab® cable S7-DIN
- Length:1,2 m
- Seq:162