Orders LinkContactUsLinkComponent
Cables
InLab® cable MultiPin-BNC/RCA (IP67), 1,8 m
Catalog # 661-0256
Supplier:   Mettler - Toledo
Cables
Catalog # 661-0256
Supplier:  Mettler - Toledo
Cables
Cables
InLab® cable MultiPin-BNC/RCA (IP67), 1,8 m
Price
156,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Specifications
  • Description:
    InLab® cable MultiPin-BNC/RCA (IP67)
  • Length:
    1,8 m
  • Seq:
    157