Cables
InLab® cable MultiPin-BNC/4mm, 1,2 m
N° de catalogue 661-0252
Fournisseur: Mettler - Toledo
Jump to:
Product Details & Documents
These electrode cables are designed to connect Mettler sensors to almost any meter.
Documents
Mettler InLab® Sensors
Product Literature
Specifications
- Description:InLab® cable MultiPin-BNC/4mm
- Length:1,2 m
- Seq:153