Order Entry
Belgium
Orders LinkContactUsLinkComponent
Microscope slides for printers

Microscope slides for printers

Purple
N° de catalogue KLINKP-SIL-3062
Fournisseur:   KLINIPATH
Microscope slides for printers
N° de catalogue KLINKP-SIL-3062
Fournisseur:  KLINIPATH
Microscope slides for printers

Purple

Prix
270,00 €
Connexion Pour la disponibilité
1 * 2.500 ST
Product Details & Documents

Microscope slides for slide printers.

  • Frosted ends
  • 90° ground edges
  • Finished clipped corners of 1 mm

Allowing for easy handling of automated slide printers and digital slide imagers in slide racks.

Spécifications
  • Colour:
    Purple
  • Seq:
    14
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.