Order Entry
Belgium
Orders LinkContactUsLinkComponent
Microscope slides for printers

Microscope slides for printers

Pink
Catalog # 631-1234
Supplier:   KLINIPATH
Microscope slides for printers
Catalog # 631-1234
Supplier:  KLINIPATH
Microscope slides for printers

Pink

Price
134,00 €
Log in for availability
1 * 2.500 ST
Product Details & Documents

Microscope slides for slide printers allowing for easy handling of automated slide printers and digital slide imagers in slide racks.

  • Frosted ends
  • 90° grounded edges
  • Finished clipped corners of 1mm
Specifications
  • Colour:
    Pink
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.