Order Entry
Belgium
Orders LinkContactUsLinkComponent
Microscope slides for printers

Microscope slides for printers

Microscope slides with frosted ends on both sides
Catalog # 631-1231
Supplier:   KLINIPATH
Microscope slides for printers
Catalog # 631-1231
Supplier:  KLINIPATH
Microscope slides for printers

Microscope slides with frosted ends on both sides

Price
76,10 €
Log in for availability
1 * 2.500 ST
Product Details & Documents

Microscope slides for slide printers allowing for easy handling of automated slide printers and digital slide imagers in slide racks.

  • Frosted ends
  • 90° grounded edges
  • Finished clipped corners of 1mm
Specifications
  • Description:
    Microscope slides with frosted ends on both sides
  • Seq:
    02