Heated forceps, eTemp
Straight forceps, 2.0 mm
Catalog # KLIN4509220
Supplier: KLINIPATH
Jump to:
Product Details & Documents
eTemp is an electrically heated forceps system that can be used in the preparation of histological or histopathological samples.
- Housing made from welded stainless steel and powder coated
- No temperature adjustment
- No tiresome reheating — full attention on processing samples
- Faster embedding increases productivity
- No contact problems at forceps connection
To ensure that small amounts of preparations do not remain on cooled forcep tips, the forceps must be repeatedly removed from the preparation and heated via external means such as a flame or an embedding station. The eTemp operates at a safe low voltage and features temperature-regulated heating elements that constantly keep the forcep tips at the optimum temperature for handling preparations in histological paraffin. This model features all-metal forceps with medical plastic insulation, which are heated exclusively at the tips.
Specifications
- Description:Straight forceps, 2.0 mm
- Seq:04