Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ethanol absolute ≥99%

Ethanol absolute ≥99%

Canister, 5 L
Catalog # VWRK4099-9005
Flammable
Irritant
Supplier:   KLINIPATH
Ethanol absolute ≥99%
Catalog # VWRK4099-9005
Supplier:  KLINIPATH
CAS Number:  64-17-5
Ethanol absolute ≥99%

Canister, 5 L

CAS Number:  64-17-5
Price
23,80 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:C₂H₅OH
  • MDL Number:MFCD00003568
  • Molecular Weight:46.07 g/mol
  • Flash Pt:12 °C (closed cup)
  • Storage Temperature:Ambient
  • UN:1170
  • ADR:3,II

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Canister
  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.