ContactUsLinkComponent
Attribute Value Port Connection: 1/4 Tube (Straight) Flow Rate Filter Rating: 230°F (110°C) at 0 psig Pressure Rating: 125 psig at 110°F Housing Material: Nylon 1 * 10 items
Catalog # PARK9933-05-BK
Supplier:  PARKER HANNIFIN
Attribute Value Port Connection: 1/4 Tube (Straight) Flow Rate Filter Rating: 230°F (110°C) at 0 psig Pressure Rating: 125 psig at 110°F Housing Material: Nylon 1 * 10 items
Catalog # PARK9933-05-BK
Supplier:  PARKER HANNIFIN
Frequently Bought Together