Temperature control systems, PRESTO®
Product Details & Documents
PRESTO® systems cover a working temperature range of −93 to +250 °C with high cooling and heating capacity. These instruments can compensate for exothermic and endothermic reactions with extraordinary speed, they feature high flow rates, constant pressure and a controlled build-up of pump pressure. Changes in the temperature control of liquid viscosity are balanced dynamically. Permanent internal monitoring and self-lubricating pumps contribute to long service life. The integrated touch panel gives the user a clear view of important information. Fingertip control via the touch screen. Multiple interfaces enable remote control of the PRESTO® across networks and in super-ordinated control systems. Wireless remote control of the new PRESTO® can easily be done by using the Wireless TEMP range of products.
- Fingertip control via the touch screen with 14,4 cm colour touch panel
- Up to three user levels with password management
- USB (Host und Device), Ethernet and SD card slot
- Units are whisper-quiet and barely audible when operating
- Filling system is accessible from the top
- Choice of models with environmentally-friendly natural refrigerant
Max. flow rate/max. pressure:
A30: 25 L/min, 0,5 bar
A38/A41/W41: 0 to 50 L/min, 0,1 to 1,6 bar
A40/W40, A80/W80: 16 to 40 L/min, 0,3 to 1,7 bar
A45/A45t/W50/W50t: 35 to 76 L/min, 0,48 to 3,2 bar
A70: 0 to 40 L/min, 0,1 to 1,7 bar W91/W92 (t/tt): 26 to 80 L/min, 0,5 to 3,0 bar W91×/W92× (t/tt): 18 to 70 L/min, 0,8 to 5,5 bar
W93: 0 to 80 L/min, 0,1 to 3 bar W93x: 0 to 70 L/min, 0,1 to 5,5 bar.
: Supplied without accessories. Go to website for details of accessories.
: Cooling capacity at +200, +100, +20, 0, –10, –20, –30 and −40 °C. W91/W92/W93 (t/tt/x): Cooling capacity at +200, +100, +20, 0, –20, –40, –60 and −80 °C.
Specifications
- Heating capacity:12 kW
- Temperature Range:–50...+250 °C
- Weight:210 kg
- Seq:012
- W×D×H:530×665×1260 mm
- Refrigerant:R452A
- Cooling capacity:7; 7; 7,5; 6,5; 5; 3; 1,8; 0,6 kW
- Temp. stability:±0,05...0,1 °C
- Type:W50t
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to M16×1 male2× adapters, M24×1,5 female to M16×1 maleJULA8890052All New Presto® series unitsLog in for availability1 * 2 ST - 170,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to tube ½"2× adapters, M24×1,5 female to tube ½"JULA8890065All New Presto® series unitsLog in for availability1 * 2 ST - 225,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to tube 1"2× adapters, M24×1,5 female to tube 1"JULA8890066All New Presto® series unitsLog in for availability1 * 2 ST - 225,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to M24×1,5 female2× adapters, M24×1,5 female to M24×1,5 femaleJULA8890067All New Presto® series unitsLog in for availability1 * 2 ST - 247,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to M30×1,5 male2× adapters, M24×1,5 female to M30×1,5 maleJULA8890068All New Presto® series unitsLog in for availability1 * 2 ST - 272,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 male to M30×1,5 female2× adapters, M24×1,5 male to M30×1,5 femaleJULA8890069All New Presto® series unitsLog in for availability1 * 2 ST - 303,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to M30×1,5 female2× adapters, M24×1,5 female to M30×1,5 femaleJULA8890070All New Presto® series unitsLog in for availability1 * 2 ST - 326,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 male to M16×1 female2× adapters, M24×1,5 male to M16×1 femaleJULA8890071All New Presto® series unitsLog in for availability1 * 2 ST - 278,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to NPT ¾" female2× adapters, M30×1,5 female to NPT ¾" femaleJULA8890084All New Presto® series unitsLog in for availability1 * 2 ST - 238,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Profibus DP interface, for instruments with RS232Profibus DP interface, for instruments with RS232JULA8900020All New Presto® series unitsLog in for availability1 * 1 ST - 1 620,00 €
Product Information
- Catalog #ForAvailabilityPrice
- RS485 interface, for instruments with RS232RS485 interface, for instruments with RS232JULA8900024All New Presto® series unitsLog in for availability1 * 1 ST - 1 620,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Pressure sensor, 2 fittings M30×1,5 male (−95 ... +250 °C)Pressure sensor, 2 fittings M30×1,5 male (−95 ... +250 °C)JULA8980772All New Presto® series unitsLog in for availability1 * 1 ST - 637,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Pressure sensor, 2 fittings M38×1,5 male (−95 ... +250 °C)Pressure sensor, 2 fittings M38×1,5 male (−95 ... +250 °C)JULA8980773All New Presto® series unitsLog in for availability1 * 1 ST - 728,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Electronic module with analog connectors (input, output, standby-in)Electronic module with analog connectors (input, output, standby-in)JULA8900105All New Presto® series unitsLog in for availability1 * 1 ST - 608,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Pressure sensor, 2 fittings M24×1,5 male (−95 ... +250 °C)Pressure sensor, 2 fittings M24×1,5 male (−95 ... +250 °C)JULA8980771All New Presto® series unitsLog in for availability1 * 1 ST - 614,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to tube ¾"2× adapters, M24×1,5 female to tube ¾"JULA8890064All New Presto® series unitsLog in for availability1 * 2 ST - 155,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to M38×1,5 male2× adapters, M30×1,5 female to M38×1,5 maleJULA8890080All New Presto® series unitsLog in for availability1 * 2 ST - 277,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 male to M38×1,5 female2× adapters, M30×1,5 male to M38×1,5 femaleJULA8890081All New Presto® series unitsLog in for availability1 * 2 ST - 277,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to NPT ¾" male2× adapters, M30×1,5 female to NPT ¾" maleJULA8890083All New Presto® series unitsLog in for availability1 * 2 ST - 269,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to NPT 1" male2× adapters, M30×1,5 female to NPT 1" maleJULA8890085All New Presto® series unitsLog in for availability1 * 2 ST - 238,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to NPT 1" female2× adapters, M30×1,5 female to NPT 1" femaleJULA8890086All New Presto® series unitsLog in for availability1 * 2 ST - 261,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to tube 1"2× adapters, M30×1,5 female to tube 1"JULA8890087All New Presto® series unitsLog in for availability1 * 2 ST - 210,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to tube ¼"2× adapters, M24×1,5 female to tube ¼"JULA8890063All New Presto® series unitsLog in for availability1 * 2 ST - 155,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to M38×1,5 female2× adapters, M30×1,5 female to M38×1,5 femaleJULA8890082All New Presto® series unitsLog in for availability1 * 2 ST - 310,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Sight glass, −100…+280 °C, PN16/Class 230, 2 fittings M30×1,5 maleSight glass, −100…+280 °C, PN16/Class 230, 2 fittings M30×1,5 maleJULA8970815All New Presto® series unitsLog in for availability1 * 1 ST - 1 610,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT 1" male2× adapters, M24×1,5 female to NPT 1" maleJULA8890062All New Presto® series unitsLog in for availability1 * 2 ST - 237,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Adapter, M24×1,5 male to M24×1,5 maleAdapter, M24×1,5 male to M24×1,5 maleJULA8890110All New Presto® series unitsLog in for availability1 * 1 ST - 90,70 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ¾" male2× adapters, M24×1,5 female to NPT ¾" maleJULA8890061All New Presto® series unitsLog in for availability1 * 2 ST - 237,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ½" male2× adapters, M24×1,5 female to NPT ½" maleJULA8890060All New Presto® series unitsLog in for availability1 * 2 ST - 237,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ³/₈" male2× adapters, M24×1,5 female to NPT ³/₈" maleJULA8890059All New Presto® series unitsLog in for availability1 * 2 ST - 161,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ¼" male2× adapters, M24×1,5 female to NPT ¼" maleJULA8890058All New Presto® series unitsLog in for availability1 * 2 ST - 152,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT 1" female2× adapters, M24×1,5 female to NPT 1" femaleJULA8890057All New Presto® series unitsLog in for availability1 * 2 ST - 303,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ¾" female2× adapters, M24×1,5 female to NPT ¾" femaleJULA8890056All New Presto® series unitsLog in for availability1 * 2 ST - 269,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ½" female2× adapters, M24×1,5 female to NPT ½" femaleJULA8890055All New Presto® series unitsLog in for availability1 * 2 ST - 240,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ³/₈" female2× adapters, M24×1,5 female to NPT ³/₈" femaleJULA8890054All New Presto® series unitsLog in for availability1 * 2 ST - 240,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M24×1,5 female to NPT ¼" female2× adapters, M24×1,5 female to NPT ¼" femaleJULA8890053All New Presto® series unitsLog in for availability1 * 2 ST - 170,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to M38×1,5 female2× adapters, M30×1,5 female to M38×1,5 femaleJULA8890089All New Presto® series unitsLog in for availability1 * 2 ST - 455,00 €
Product Information
- Catalog #ForAvailabilityPrice
- 2× adapters, M30×1,5 female to M30×1,5 female2× adapters, M30×1,5 female to M30×1,5 femaleJULA8890088All New Presto® series unitsLog in for availability1 * 2 ST - 334,00 €
Product Information
- Catalog #ForAvailabilityPrice
- EtherCAT interface is a protocol converter from EtherCAT to RS232EtherCAT interface is a protocol converter from EtherCAT to RS232JULA8900025All New Presto® series unitsLog in for availability1 * 1 ST - 1 620,00 €