ContactUsLinkComponent
300µl Pre-Sterilized Hamilton CO-RE Style Clear Filter Tip. 96 Tips per Rack, 5 Racks/pack, 12 packs/case 1 * 5.760 items
Catalog # AXYGHTF-300-CHTRS5
Supplier:  Corning
300µl Pre-Sterilized Hamilton CO-RE Style Clear Filter Tip. 96 Tips per Rack, 5 Racks/pack, 12 packs/case 1 * 5.760 items
Catalog # AXYGHTF-300-CHTRS5
Supplier:  Corning
Frequently Bought Together