Orders LinkContactUsLinkComponent
Objectives for CKX53
Objectives for CKX53
Dry objective with pre-centre phase contrast slider, 8,8 mm
Catalog # OLYMN5229200
Supplier:   OLYMPUS OPTICAL
Objectives for CKX53
Catalog # OLYMN5229200
Supplier:  OLYMPUS OPTICAL
Objectives for CKX53
Objectives for CKX53
Dry objective with pre-centre phase contrast slider, 8,8 mm
Price
221,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    Ph 10×/0,25
  • Description:
    Dry objective with pre-centre phase contrast slider
  • Seq:
    021
  • Working distance:
    8,8 mm