Order Entry
Belgium
Orders LinkContactUsLinkComponent
Electrolyte solutions

Electrolyte solutions

Electrolyte solution (KCl 3M), 125 ml, 125 mL
Catalog # HACHLZW9510.99
Supplier:   Hach
Electrolyte solutions
Catalog # HACHLZW9510.99
Supplier:  Hach
CAS Number:  
Electrolyte solutions
Electrolyte solutions

Electrolyte solution (KCl 3M), 125 ml, 125 mL

CAS Number:  
Price
19,00 €
Log in for availability
1 * 125 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    125 mL
  • Description:
    Electrolyte solution (KCl 3M), 125 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.