Orders LinkContactUsLinkComponent
Modular storage systems, KP
Modular storage systems, KP
KP-BLOCK, modular and stackable block storage metal cabinet with 13 drawers, yellow - capacity: 700 blocks
Catalog # KLIN252000-Y
Supplier:   KLINIPATH
Modular storage systems, KP
Catalog # KLIN252000-Y
Supplier:  KLINIPATH
Modular storage systems, KP
Modular storage systems, KP
KP-BLOCK, modular and stackable block storage metal cabinet with 13 drawers, yellow - capacity: 700 blocks
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Stackable, universal storage system for preparations and blocks. A complete system with growth potential. A modular system able to transform local storage in a laboratory into a long-term storage system. A growth system with easy exchangeable and removable plastic trays.
  • Lower gliding resistance, no rusting because of paint damage
Specifications
  • Description:
    KP-BLOCK, modular and stackable block storage metal cabinet with 13 drawers, yellow - capacity: 700 blocks
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Top for modular systems, KP Glass and KP Block
    Top for modular systems, KP Glass and KP Block
    720-2813
    Log in for availability
    1 * 1 ST - 9,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Base for modular systems, KP Glass and KP Block
    Base for modular systems, KP Glass and KP Block
    720-2812
    Log in for availability
    1 * 1 ST - 62,90 €