Orders LinkContactUsLinkComponent
Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
Orange
Catalog # KLIN110-06T
Supplier:   KLINIPATH
Embedding cassettes with disposable lid
Catalog # KLIN110-06T
Supplier:  KLINIPATH
Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
Orange
Price
163,00 €
Log in for availability
1 * 1.600 ST
Product Details & Documents
Embedded cassettes with hinged lid.
  • Cassettes maintain their stability, rigidness and shape during processing of tissues
Informations de commande: Available in different colours.
Emballage: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
Specifications
  • Colour:
    Orange
  • Seq:
    30