Orders LinkContactUsLinkComponent
Single-use bags for HYPERStack® vessels
Single-use bags for HYPERStack® vessels
Media bag, 20 L
Catalog # MDTC91-200-77
Supplier:   Corning
Single-use bags for HYPERStack® vessels
Catalog # MDTC91-200-77
Supplier:  Corning
Single-use bags for HYPERStack® vessels
Media bag, 20 L
Price
194,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors.
They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
Specifications
  • Capacity:
    20 L
  • Description:
    Media bag
  • Seq:
    03
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.