Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Spill control pallets, EcoPolyBlend™

Spill control pallet, black, 276 liter sump capacity, for 4 square drums, load capacity 2268 kg, 32,0 kg, 1245×1245×260 mm
Catalog # 119-0378
Spill control pallets, EcoPolyBlend™
Catalog # 119-0378
Spill control pallets, EcoPolyBlend™
Spill control pallets, EcoPolyBlend™

Spill control pallet, black, 276 liter sump capacity, for 4 square drums, load capacity 2268 kg, 32,0 kg, 1245×1245×260 mm

Price
541,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These pallets are made of up to 100% recycled polyethylene; dirt hiding black colour is 100% recycled, high visibility yellow is 35% recycled content. Fully compliant and quality engineered with a robust design, they withstand the toughest environments and are nearly impervious to chemical attack.

  • Independently validated by UL Environment
  • Seamless, robust design eliminates leaks
  • Excellent chemical compatibility and no corrosion
  • Sturdy and self-locating grates remove for easy cleaning
  • Square or space saving linear styles in multiple sizes offer storage versatility

 : Meets or exceeds EPA, SPCC, NFPA 1, and the Int’l Fire Code.

Specifications
  • Weight:
    32,0 kg
  • Description:
    Spill control pallet, black, 276 liter sump capacity, for 4 square drums, load capacity 2268 kg
  • Seq:
    03
  • W×D×H:
    1245×1245×260 mm
Accessories

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Ramp, for 4 drum square
    Ramp, for 4 drum square
    119-0391
    1245 x 838 x 267 mm
    Log in for availability
    1 * 1 ST - 492,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.